Gene Transcripts

Gene Symbol
ENKUR
Entrez Gene ID
219670
Gene Name
enkurin, TRPC channel interacting protein
Gene Aliases
C10orf63, CFAP106
Chromosome Location
Chr. 10: 24981979 - 25062279 on Build GRCh38
UniGene ID
Hs.534486
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270383.1 NP_001257312.1 3, 4 588
NM_145010.3 NP_659447.1 3, 4 675
NR_072992.1 3, 4 675
XM_006717389.1 XP_006717452.1 3, 4 671
XM_006717390.1 XP_006717453.1 2, 3 494
XM_011519348.1 XP_011517650.1 3, 4 671
XM_017015830.1 XP_016871319.1 2, 3 494
GenBank mRNA AK095021.1 - 3, 4 620
AK292495.1 - 3, 4 672
AY454125.1 - 3, 4 662
BC026165.2 - 3, 4 677 4837530
CR936720.1 - 3, 4 588

View Full Product Details