Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001177800.1 | NP_001171271.1 | 4 | 798 | |
NM_004797.3 | NP_004788.1 | 3 | 747 | ||
GenBank mRNA | AK291525.1 | - | 3 | 720 | |
AK312868.1 | - | 4 | 768 | ||
AX767959.1 | - | 3 | 689 | ||
BC054496.1 | - | 688 | 6192794 | ||
BC096308.3 | - | 3 | 731 | 40007540 | |
BC096309.3 | - | 3 | 731 | 40007541 | |
BC096310.3 | - | 3 | 731 | 40007542 | |
BC096311.3 | - | 3 | 731 | 40007544 | |
D45371.1 | - | 3 | 689 | ||
EU420013.1 | - | 2 | 663 |