Gene Transcripts

Gene Symbol
CHRDL2
Entrez Gene ID
25884
Gene Name
chordin like 2
Gene Aliases
BNF1, CHL2
Chromosome Location
Chr. 11: 74696428 - 74731385 on Build GRCh38
UniGene ID
Hs.432379
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278473.2 NP_001265402.1 11 1785
NM_001304390.1 NP_001291319.1 11 1732
NM_001304391.1 NP_001291320.1 10 1537
NM_001304415.1 NP_001291344.1 9 1482
NM_001304416.1 NP_001291345.1 9 1583
NM_001304417.1 NP_001291346.1 9 1430
NM_015424.5 NP_056239.3 12 1840
GenBank mRNA AF332891.1 - 7 964
AL047873.1 - 4 325
AL110168.1 - 4 585
AY163868.1 - 12 1346
AY279090.1 - 11 1582
AY279092.1 - 9 1583
AY358522.1 - 12 1629
BC142623.1 - 11 1785 40147467
JN579714.1 - 5 398

View Full Product Details