Gene Transcripts
- Gene Symbol
- FGFR1OP2
- Entrez Gene ID
- 26127
- Gene Name
- FGFR1 oncogene partner 2
- Gene Aliases
- HSPC123-like, WIT3.0
- Chromosome Location
- Chr. 12: 26938372 - 26966648 on Build GRCh38
- UniGene ID
- Hs.591162
- Species
- Homo sapiens
- Gene Symbol
- LOC100335030
- Entrez Gene ID
- 100335030
- Gene Name
- FGFR1 oncogene partner 2 pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 12: 49389174 - 49392335 on Build GRCh38
- UniGene ID
- Hs.687044
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033267.1 |
|
1 |
542 |
|
View Full Product Details