Gene Transcripts

Gene Symbol
RBM19
Entrez Gene ID
9904
Gene Name
RNA binding motif protein 19
Gene Aliases
Chromosome Location
Chr. 12: 113816738 - 113966371 on Build GRCh38
UniGene ID
Hs.7482
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146698.1 NP_001140170.1 8 1107
NM_001146699.1 NP_001140171.1 8 1107
NM_016196.3 NP_057280.2 8 1107
XM_017020280.1 XP_016875769.1 8 1107
XM_017020281.1 XP_016875770.1 8 1107
GenBank mRNA AB014582.1 - 8 1042
AK291444.1 - 8 1058
AK314606.1 - 8 1107
AL117547.1 - 8 1048
BC004289.1 - 8 1046 3619715
BC006137.1 - 8 1046 3677625

View Full Product Details