Gene Transcripts

Gene Symbol
MIER2
Entrez Gene ID
54531
Gene Name
MIER family member 2
Gene Aliases
KIAA1193, Mi-er2
Chromosome Location
Chr. 19: 305573 - 344793 on Build GRCh38
UniGene ID
Hs.101891
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017550.1 NP_060020.1 14 2367
XM_005259582.4 XP_005259639.1 14 2951
XM_006722769.3 XP_006722832.1 14 2934
XM_011528072.2 XP_011526374.1 14 2968
XM_011528073.1 XP_011526375.1 14 2384
XM_011528074.2 XP_011526376.1 14 2951
XM_011528075.2 XP_011526377.1 14 3489
XM_011528077.2 XP_011526379.1 15 5330
XM_011528081.2 XP_011526383.1 12 2226
XM_011528082.2 XP_011526384.1 12 2157
GenBank mRNA AB033019.1 - 2355
BC028203.1 - 14 2367 5201526

View Full Product Details