Gene Transcripts
- Gene Symbol
- GGNBP2
- Entrez Gene ID
- 79893
- Gene Name
- gametogenetin binding protein 2
- Gene Aliases
- DIF-3, DIF3, LCRG1, LZK1, ZFP403, ZNF403
- Chromosome Location
- Chr. 17: 36544888 - 36589848 on Build GRCh38
- UniGene ID
- Hs.514116
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_024835.4 |
NP_079111.1
|
3, 4 |
498 |
|
XM_005257689.2 |
XP_005257746.1
|
3, 4 |
474 |
|
XM_011525265.2 |
XP_011523567.1
|
2, 3 |
237 |
|
XM_011525266.1 |
XP_011523568.1
|
3, 4 |
474 |
|
XM_017025107.1 |
XP_016880596.1
|
2, 3 |
211 |
|
GenBank mRNA |
AF126964.1 |
- |
2, 3 |
229 |
|
AF268387.1 |
- |
3, 4 |
418 |
|
AK024883.1 |
- |
3, 4 |
454 |
|
AK290687.1 |
- |
2, 3 |
254 |
|
AK292161.1 |
- |
3, 4 |
467 |
|
BC137491.1 |
- |
3, 4 |
364 |
9021491 |
BG393068.1 |
- |
|
431 |
|
DA494492.1 |
- |
3, 4 |
499 |
|
DC424145.1 |
- |
3, 4 |
474 |
|
View Full Product Details