Gene Transcripts

Gene Symbol
MOSPD2
Entrez Gene ID
158747
Gene Name
motile sperm domain containing 2
Gene Aliases
Chromosome Location
Chr. X: 14873405 - 14922166 on Build GRCh38
UniGene ID
Hs.744919
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177475.1 NP_001170946.1 14 2821
NM_152581.3 NP_689794.1 15 2857
GenBank mRNA AL834345.1 - 7 1909
BC030641.1 - 15 2824 4821861

View Full Product Details