Gene Transcripts

Gene Symbol
RPRML
Entrez Gene ID
388394
Gene Name
reprimo like
Gene Aliases
Chromosome Location
Chr. 17: 46978156 - 46979248 on Build GRCh38
UniGene ID
Hs.367999
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_203400.4 NP_981945.1 1 1025
GenBank mRNA BC033942.1 - 1 1032
BC108690.1 - 1 1002 6140781

View Full Product Details