Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009307.3 | NP_033333.2 | 10 | 1506 | |
GenBank mRNA | AF257304.1 | - | 9 | 1261 | |
AK036357.1 | - | 10 | 1507 | ||
AK220537.1 | - | 7 | 820 | ||
BC027019.1 | - | 9 | 1267 | 5362352 | |
D37793.1 | - | 8 | 1140 |