Gene Transcripts

Gene Symbol
Serf1
Entrez Gene ID
20365
Gene Name
small EDRK-rich factor 1
Gene Aliases
4F5, Msmac1, m4F5
Chromosome Location
Chr. 13: 100108019 - 100114233 on Build GRCm38
UniGene ID
Mm.286177
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011353.2 NP_035483.1 3 421
GenBank mRNA AF073517.1 - 3 392
AK005773.1 - 3 336
BC028510.1 - 3 374 1510984

View Full Product Details