Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011374.2 | NP_035504.2 | 4, 5 | 1258 | |
GenBank mRNA | AK042207.1 | - | 4, 5 | 1114 | |
AK046188.1 | - | 4, 5 | 1198 | ||
AK052444.1 | - | 4, 5 | 909 | ||
BC024821.1 | - | 4, 5 | 1258 | 5357151 | |
EF101558.1 | - | 4, 5 | 370 | ||
L38677.1 | - | 4, 5 | 882 | ||
X84235.1 | - | 4, 5 | 605 |