Gene Transcripts

Gene Symbol
Oas2
Entrez Gene ID
246728
Gene Name
2'-5' oligoadenylate synthetase 2
Gene Aliases
Oasl11
Chromosome Location
Chr. 5: 120730333 - 120749884 on Build GRCm38
UniGene ID
Mm.260926
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145227.3 NP_660262.2 11 3191
GenBank mRNA AB067535.1 - 11 3196
AF418010.1 - 12 3225
AK049218.1 - 3 1705

View Full Product Details