Gene Transcripts

Gene Symbol
Ddx26b
Entrez Gene ID
236790
Gene Name
DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 26B
Gene Aliases
4930535D10Rik, 6330505F04Rik, D130066O12
Chromosome Location
Chr. X: 56454821 - 56507843 on Build GRCm38
UniGene ID
Mm.474772
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172779.4 NP_766367.1 3 576
GenBank mRNA AK031945.1 - 3 568
AK083942.1 - 3 538
AK160627.1 - 3 536
BC051161.1 - 3 558 6466142

View Full Product Details