Gene Transcripts

Gene Symbol
Dpp10
Entrez Gene ID
269109
Gene Name
dipeptidylpeptidase 10
Gene Aliases
6430601K09Rik, DPP X, Dprp3
Chromosome Location
Chr. 1: 123332138 - 124845960 on Build GRCm38
UniGene ID
Mm.417716
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199021.3 NP_950186.3 10 998
GenBank mRNA AK032579.1 - 8 644
AK046842.1 - 10 999
BC067026.1 - 10 991 6853947

View Full Product Details