Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008720.2 | NP_032746.2 | 4 | 487 | |
GenBank mRNA | AF003348.1 | - | 4 | 464 | |
AK044878.1 | - | 3 | 2879 | ||
AK087174.1 | - | 3 | 2896 | ||
AK154282.1 | - | 4 | 407 | ||
AK155379.1 | - | 4 | 407 | ||
AK170880.1 | - | 3 | 2880 | ||
AK171371.1 | - | 2 | 3998 | ||
BC052437.1 | - | 4 | 487 | 6405214 | |
BC054539.1 | - | 4 | 487 | 6405214 |