Gene Transcripts

Gene Symbol
Ly86
Entrez Gene ID
17084
Gene Name
lymphocyte antigen 86
Gene Aliases
MD-1, MD1
Chromosome Location
Chr. 13: 37345345 - 37419036 on Build GRCm38
UniGene ID
Mm.2639
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010745.2 NP_034875.1 2 282
GenBank mRNA AB007599.1 - 2 282
AK027929.1 - 2 271
BC065783.1 - 2 187
BC096414.1 - 2 250 849991

View Full Product Details