Gene Transcripts
- Gene Symbol
- Mpdz
- Entrez Gene ID
- 17475
- Gene Name
- multiple PDZ domain protein
- Gene Aliases
- AI225843, B930003D11Rik, MUPP1
- Chromosome Location
- Chr. 4: 81278499 - 81442918 on Build GRCm38
- UniGene ID
- Mm.153039
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF326526.1 |
- |
9, 10 |
1289 |
|
AF326527.1 |
- |
9, 10 |
1289 |
|
AF326528.1 |
- |
9, 10 |
1289 |
|
AF326529.1 |
- |
9, 10 |
1289 |
|
AF326530.1 |
- |
9, 10 |
1289 |
|
AF326531.2 |
- |
9, 10 |
1289 |
|
AF326532.1 |
- |
9, 10 |
1289 |
|
AF326533.1 |
- |
9, 10 |
1289 |
|
AF326534.1 |
- |
9, 10 |
1289 |
|
AF326535.1 |
- |
9, 10 |
1289 |
|
AF326536.1 |
- |
9, 10 |
1289 |
|
AF326537.1 |
- |
9, 10 |
1289 |
|
AF326538.1 |
- |
9, 10 |
1289 |
|
AF326539.1 |
- |
9, 10 |
1289 |
|
AF326540.1 |
- |
9, 10 |
1289 |
|
AF326541.1 |
- |
9, 10 |
1289 |
|
AF326542.1 |
- |
9, 10 |
1289 |
|
AF326543.1 |
- |
9, 10 |
1289 |
|
AF326544.1 |
- |
9, 10 |
1289 |
|
AJ131869.1 |
- |
10, 11 |
1380 |
|
BC138045.1 |
- |
9, 10 |
1352 |
8861064 |
BC145115.1 |
- |
9, 10 |
1352 |
|
BC145117.1 |
- |
9, 10 |
1352 |
9053665 |
View Full Product Details