Gene Transcripts

Gene Symbol
C21orf2
Entrez Gene ID
755
Gene Name
chromosome 21 open reading frame 2
Gene Aliases
LRRC76, YF5/A2
Chromosome Location
Chr. 21: 44328944 - 44339417 on Build GRCh38
UniGene ID
Hs.517331
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271440.1 NP_001258369.1 7 1799
NM_001271441.1 NP_001258370.1 7 2159
NM_001271442.1 NP_001258371.1 8 1707
NM_004928.2 NP_004919.1 7 1802
XM_006724051.3 XP_006724114.1 6 1931
XM_006724053.3 XP_006724116.1 6 1608
XM_017028470.1 XP_016883959.1 7 1810
XM_017028471.1 XP_016883960.1 6 1632
XM_017028472.1 XP_016883961.1 6 1687
GenBank mRNA Y11392.1 - 7 1662
Z93322.1 - 7 1601

View Full Product Details