Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013566.2 | NP_038594.2 | 7 | 1114 | |
GenBank mRNA | AK078871.1 | - | 7 | 1115 | |
AK144773.1 | - | 7 | 1087 | ||
AK154766.1 | - | 7 | 1115 | ||
AK155207.1 | - | 7 | 1081 | ||
AK155594.1 | - | 7 | 1087 | ||
BC011184.1 | - | 7 | 1087 | 4187781 | |
M68903.1 | - | 7 | 1062 | ||
M95632.1 | - | 7 | 1058 | ||
M95633.1 | - | 7 | 1058 | ||
S44607.1 | - | 6 | 1026 |