Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013622.3 | NP_038650.3 | 3 | 1765 | |
GenBank mRNA | AK043873.1 | - | 3 | 1766 | |
BC137958.1 | - | 3 | 1691 | 8860977 | |
BC137969.1 | - | 3 | 1692 | 8860988 | |
L06322.1 | - | 3 | 1722 | ||
L07271.1 | - | 3 | 1692 | ||
L11064.1 | - | 3 | 1663 | ||
S65335.1 | - | 3 | 1691 | ||
S66181.1 | - | 3 | 1643 |