Gene Transcripts
- Gene Symbol
- PDE1C
- Entrez Gene ID
- 5137
- Gene Name
- phosphodiesterase 1C
- Gene Aliases
- Hcam3, cam-PDE 1C, hCam-3
- Chromosome Location
- Chr. 7: 31616777 - 32428224 on Build GRCh38
- UniGene ID
- Hs.143821
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001191056.3 |
NP_001177985.1
|
3, 4 |
719 |
|
NM_001191057.3 |
NP_001177986.1
|
3, 4 |
719 |
|
NM_001191058.3 |
NP_001177987.2
|
4, 5 |
1099 |
|
NM_001191059.3 |
NP_001177988.1
|
4, 5 |
710 |
|
NM_001322055.1 |
NP_001308984.1
|
4, 5 |
945 |
|
NM_001322056.1 |
NP_001308985.1
|
4, 5 |
710 |
|
NM_001322057.1 |
NP_001308986.1
|
4, 5 |
945 |
|
NM_001322058.1 |
NP_001308987.1
|
4, 5 |
1099 |
|
NM_001322059.1 |
NP_001308988.1
|
4, 5 |
748 |
|
NM_005020.4 |
NP_005011.1
|
4, 5 |
483 |
|
XM_017012264.1 |
XP_016867753.1
|
4, 5 |
550 |
|
XM_017012265.1 |
XP_016867754.1
|
4, 5 |
401 |
|
XM_017012266.1 |
XP_016867755.1
|
4, 5 |
550 |
|
XM_017012267.1 |
XP_016867756.1
|
4, 5 |
455 |
|
GenBank mRNA |
AK056170.1 |
- |
3, 4 |
706 |
|
AK091734.1 |
- |
4, 5 |
1024 |
|
AK094180.1 |
- |
4, 5 |
642 |
|
BC022479.1 |
- |
3, 4 |
720 |
4795813 |
BC022525.1 |
- |
3, 4 |
720 |
4795158 |
DA025688.1 |
- |
4, 5 |
466 |
|
DA174241.1 |
- |
4, 5 |
507 |
|
U40371.1 |
- |
4, 5 |
426 |
|
View Full Product Details