Gene Transcripts

Gene Symbol
Gapdhs
Entrez Gene ID
14447
Gene Name
glyceraldehyde-3-phosphate dehydrogenase, spermatogenic
Gene Aliases
Gapd-s, Gapds
Chromosome Location
Chr. 7: 30729777 - 30744791 on Build GRCm38
UniGene ID
Mm.436562
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290631.1 NP_001277560.1 4 521
NM_008085.2 NP_032111.1 4 539
GenBank mRNA M60978.1 - 4 521

View Full Product Details