Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008138.4 | NP_032164.2 | 8 | 1060 | |
GenBank mRNA | AK157998.1 | - | 8 | 1064 | |
AK159222.1 | - | 8 | 1064 | ||
AK160186.1 | - | 8 | 1057 | ||
AK167388.1 | - | 8 | 1062 | ||
AK179809.1 | - | 2 | 209 | ||
AK212898.1 | - | 2 | 113 | ||
BC006695.1 | - | 1 | 31 | ||
BC037130.1 | - | 4 | 384 | ||
BC065159.1 | - | 8 | 1047 | 5717305 | |
M13963.1 | - | 8 | 1032 |