Gene Transcripts

Gene Symbol
Htr4
Entrez Gene ID
15562
Gene Name
5 hydroxytryptamine (serotonin) receptor 4
Gene Aliases
5-HT-4, 5-HT4, 5-HT<4L>, 5HTR4
Chromosome Location
Chr. 18: 62322160 - 62498026 on Build GRCm38
UniGene ID
Mm.20440
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008313.4 NP_032339.2 4 536
GenBank mRNA AJ011369.1 - 3 326
AK141587.1 - 4 480
AK141625.1 - 4 537
BC132189.1 - 3 365
BC132191.1 - 3 365
BC145159.1 - 3 365
BC145161.1 - 3 365
JN418927.1 - 4 467
JN418928.1 - 4 453
JN418929.1 - 3 326
JN418930.1 - 3 342
Y09585.1 - 3 326
Y09587.1 - 3 326
Y09588.1 - 3 326

View Full Product Details