Gene Transcripts

Gene Symbol
Nr2f1
Entrez Gene ID
13865
Gene Name
nuclear receptor subfamily 2, group F, member 1
Gene Aliases
COUP-TF1, COUP-TFI, COUPTFA, EAR3, Erbal3, SVP44, Tcfcoup1
Chromosome Location
Chr. 13: 78188971 - 78199172 on Build GRCm38
UniGene ID
Mm.439653
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010151.2 NP_034281.2 3 2018
GenBank mRNA AK148880.1 - 3 2357
X74134.1 - 3 1910

View Full Product Details