Gene Transcripts

Gene Symbol
Ccr6
Entrez Gene ID
12458
Gene Name
chemokine (C-C motif) receptor 6
Gene Aliases
CC-CKR-6, CCR-6, Cmkbr6, KY411
Chromosome Location
Chr. 17: 8236043 - 8257141 on Build GRCm38
UniGene ID
Mm.8007
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001190333.1 NP_001177262.1 3 741
NM_001190334.1 NP_001177263.1 4 613
NM_001190335.1 NP_001177264.1 3 522
NM_001190336.1 NP_001177265.1 3 498
NM_001190337.1 NP_001177266.1 2 407
NM_001190338.1 NP_001177267.1 2 494
NM_009835.4 NP_033965.1 6 943
GenBank mRNA AB009369.1 - 2 497
AB016031.1 - 3 572
AK089712.1 - 6 944
AK154413.1 - 5 1316
BC100756.1 - 4 609 40046797
BC100757.1 - 3 518 40046796
BC100758.1 - 3 494 40046794
BC105669.1 - 2 400 40046795

View Full Product Details