Gene Transcripts

Gene Symbol
Ccr2
Entrez Gene ID
12772
Gene Name
chemokine (C-C motif) receptor 2
Gene Aliases
Cc-ckr-2, Ccr2a, Ccr2b, Ckr2, Ckr2a, Ckr2b, Cmkbr2, mJe-r
Chromosome Location
Chr. 9: 124101918 - 124109140 on Build GRCm38
UniGene ID
Mm.6272
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009915.2 NP_034045.1 3 242
GenBank mRNA AK046579.1 - 476
AK138599.1 - 2 555
AK154590.1 - 2 1991
BC138803.1 - 2 202 8861825
BC138804.1 - 2 202 8861826
U51717.1 - 2 200
U56819.1 - 3 242

View Full Product Details