Gene Transcripts

Gene Symbol
Csf3
Entrez Gene ID
12985
Gene Name
colony stimulating factor 3 (granulocyte)
Gene Aliases
Csfg, G-CSF, MGI-IG
Chromosome Location
Chr. 11: 98701095 - 98704836 on Build GRCm38
UniGene ID
Mm.1238
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009971.1 NP_034101.1 5 1148
GenBank mRNA AK145177.1 - 5 1113
M13926.1 - 5 1148

View Full Product Details