Gene Transcripts

Gene Symbol
Dnase2a
Entrez Gene ID
13423
Gene Name
deoxyribonuclease II alpha
Gene Aliases
Dnase2
Chromosome Location
Chr. 8: 84908624 - 84911461 on Build GRCm38
UniGene ID
Mm.350831
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010062.3 NP_034192.1 3 454
GenBank mRNA AK145192.1 - 3 455
AK167277.1 - 3 420
BC058609.1 - 3 401 5397016

View Full Product Details