Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_198408.3 | NP_940800.1 | 7 | 1035 | |
GenBank mRNA | AK135067.1 | - | 7 | 1036 | |
AK139404.1 | - | 7 | 881 | ||
AK164173.1 | - | 1 | 45 | ||
BC061247.1 | - | 7 | 976 | 30246702 |