Gene Transcripts
- Gene Symbol
- Anapc1
- Entrez Gene ID
- 17222
- Gene Name
- anaphase promoting complex subunit 1
- Gene Aliases
- 2610021O03Rik, AI047775, AI853536, AW547281, Apc1, Mcpr, tsg24
- Chromosome Location
- Chr. 2: 128610083 - 128687410 on Build GRCm38
- UniGene ID
- Mm.277408
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_008569.2 |
NP_032595.2
|
8, 9 |
1093 |
|
GenBank mRNA |
AK052404.1 |
- |
8, 9 |
1074 |
|
AK054305.1 |
- |
8, 9 |
1075 |
|
AK077847.1 |
- |
8, 9 |
1094 |
|
AK090134.1 |
- |
8, 9 |
1076 |
|
AK149762.1 |
- |
8, 9 |
1065 |
|
AK164804.1 |
- |
8, 9 |
1079 |
|
BC139002.1 |
- |
8, 9 |
1074 |
8862024 |
BC145492.1 |
- |
8, 9 |
1074 |
9054055 |
X80169.1 |
- |
8, 9 |
1078 |
|
View Full Product Details