Gene Transcripts

Gene Symbol
Ppp3ca
Entrez Gene ID
19055
Gene Name
protein phosphatase 3, catalytic subunit, alpha isoform
Gene Aliases
2900074D19Rik, CN, Caln, Calna, CnA
Chromosome Location
Chr. 3: 136670066 - 136937727 on Build GRCm38
UniGene ID
Mm.331389
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001293622.1 NP_001280551.1 13 2435
NM_008913.5 NP_032939.1 14 2465
GenBank mRNA AK076570.1 - 5 754
AK145309.1 - 1 57
AK146387.1 - 13 2381
AK150393.1 - 13 1794
AK166287.1 - 13 2213
AK179617.1 - 1 126
AK218972.1 - 1 125
BC024843.1 - 5 585
BC096652.1 - 6 820
J04134.1 - 9 1046
J05479.1 - 14 1763

View Full Product Details