Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009658.3 | NP_033788.3 | 5 | 612 | |
GenBank mRNA | AK028079.1 | - | 5 | 567 | |
AK138000.1 | - | 4 | 854 | ||
AK139250.1 | - | 5 | 567 | ||
AK149861.1 | - | 5 | 602 | ||
AK150197.1 | - | 5 | 567 | ||
AK153949.1 | - | 5 | 596 | ||
AK164682.1 | - | 5 | 567 | ||
AK170663.1 | - | 5 | 614 | ||
BC004725.1 | - | 5 | 559 | 3582788 | |
BC021655.1 | - | 5 | 605 | 3708927 | |
BC085310.1 | - | 5 | 544 | 6485416 | |
D32250.1 | - | 5 | 553 | ||
L39795.1 | - | 5 | 544 | ||
U29152.1 | - | 5 | 563 |