Gene Transcripts

Gene Symbol
Creb3l1
Entrez Gene ID
26427
Gene Name
cAMP responsive element binding protein 3-like 1
Gene Aliases
Oasis
Chromosome Location
Chr. 2: 91982328 - 92024170 on Build GRCm38
UniGene ID
Mm.10353
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011957.2 NP_036087.2 11 1344
GenBank mRNA AB017614.1 - 11 1306
AK142368.1 - 1345
BC016447.1 - 11 1319 4921584

View Full Product Details