Gene Transcripts
- Gene Symbol
- Ostf1
- Entrez Gene ID
- 20409
- Gene Name
- osteoclast stimulating factor 1
- Gene Aliases
- C78236, SH3P2, Sh3d3
- Chromosome Location
- Chr. 19: 18580364 - 18631813 on Build GRCm38
- UniGene ID
- Mm.172222
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017375.3 |
NP_059071.1
|
7, 8 |
570 |
|
GenBank mRNA |
AK002899.1 |
- |
7, 8 |
549 |
|
AK005525.1 |
- |
7, 8 |
539 |
|
AK008123.1 |
- |
7, 8 |
518 |
|
AK008467.1 |
- |
7, 8 |
518 |
|
AK010074.1 |
- |
7, 8 |
566 |
|
AK145949.1 |
- |
7, 8 |
521 |
|
AK149175.1 |
- |
|
548 |
|
AK150145.1 |
- |
7, 8 |
540 |
|
AK155028.1 |
- |
7, 8 |
540 |
|
BC060986.1 |
- |
7, 8 |
449 |
30300544 |
U58888.1 |
- |
7, 8 |
518 |
|
View Full Product Details