Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001205043.1 | NP_001191972.1 | 18 | 4325 | |
NM_001205044.1 | NP_001191973.1 | 19 | 3998 | ||
NM_021878.3 | NP_068678.1 | 19 | 4044 | ||
GenBank mRNA | AK045214.1 | - | 18 | 4130 | |
AK147226.1 | - | 18 | 4325 | ||
BC003374.1 | - | 12 | 1894 | ||
BC026056.1 | - | 18 | 4232 | ||
BC050129.1 | - | 18 | 4246 | ||
BC051010.1 | - | 16 | 6686 | ||
BC052444.1 | - | 19 | 3915 | 6406875 | |
BC060695.1 | - | 19 | 3915 | 6406875 | |
D31967.1 | - | 19 | 3934 |