Gene Transcripts

Gene Symbol
Fndc4
Entrez Gene ID
64339
Gene Name
fibronectin type III domain containing 4
Gene Aliases
2810430J06Rik, 6330410H20Rik, AB030187, AI838506, AW487863, FRCP1, Fnmp1
Chromosome Location
Chr. 5: 31292245 - 31295877 on Build GRCm38
UniGene ID
Mm.292544
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022424.5 NP_071869.2 7 1060
GenBank mRNA AB030187.1 - 3 232
AK013203.1 - 7 1054
AK018154.1 - 7 1056
AK163183.1 - 7 1061
BC027164.1 - 3 214
BC139278.1 - 7 812 8862300
BC139292.1 - 7 812 8862314

View Full Product Details