Gene Transcripts

Gene Symbol
Cdca7
Entrez Gene ID
66953
Gene Name
cell division cycle associated 7
Gene Aliases
2310021G01Rik, AU044109, JPO1
Chromosome Location
Chr. 2: 72476189 - 72486892 on Build GRCm38
UniGene ID
Mm.270676
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025866.3 NP_080142.1 5 624
GenBank mRNA AK011289.1 - 5 625
AK028671.1 - 5 635
AK075882.1 - 5 589
AK153943.1 - 5 603
AK167734.1 - 5 625
BC066169.1 - 5 632 30474357

View Full Product Details