Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026156.2 | NP_080432.1 | 4 | 395 | |
GenBank mRNA | AK002890.1 | - | 4 | 396 | |
AK031533.1 | - | 405 | |||
AK087952.1 | - | 4 | 402 | ||
AK132434.1 | - | 4 | 409 | ||
AK145925.1 | - | 4 | 397 | ||
AK199409.1 | - | 2 | 143 | ||
AK214621.1 | - | 2 | 143 | ||
BC021341.1 | - | 4 | 393 | 5066146 |