Gene Transcripts

Gene Symbol
Sec62
Entrez Gene ID
69276
Gene Name
SEC62 homolog (S. cerevisiae)
Gene Aliases
3100002M17Rik, AI844545, AW545092, Dtrp1, HTP1, Tloc1
Chromosome Location
Chr. 3: 30792774 - 30821263 on Build GRCm38
UniGene ID
Mm.26017
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027016.2 NP_081292.1 6 1050
GenBank mRNA AK088093.1 - 6 599
AK090078.1 - 6 600
BC023720.1 - 3 265
BC053504.1 - 6 588
BC067202.1 - 6 585 6332701
BC071243.1 - 6 601

View Full Product Details