Gene Transcripts

Gene Symbol
Slc39a11
Entrez Gene ID
69806
Gene Name
solute carrier family 39 (metal ion transporter), member 11
Gene Aliases
1810074D23Rik
Chromosome Location
Chr. 11: 113244851 - 113650752 on Build GRCm38
UniGene ID
Mm.341021
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC004643.1 - 2223
BC019647.1 - 2637 3978277
BC031751.1 - 2751 4216487

View Full Product Details