Gene Transcripts

Gene Symbol
Sertad3
Entrez Gene ID
170742
Gene Name
SERTA domain containing 3
Gene Aliases
AW124805, D1Mgi51, Rbt1, Sei3
Chromosome Location
Chr. 7: 27473840 - 27477364 on Build GRCm38
UniGene ID
Mm.200120
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133210.2 NP_573473.1 2 211
GenBank mRNA AF317202.1 - 2 215
AK134829.1 - 2 284
BC034886.1 - 2 211 1364881

View Full Product Details