Gene Transcripts

Gene Symbol
Slc16a13
Entrez Gene ID
69309
Gene Name
solute carrier family 16 (monocarboxylic acid transporters), member 13
Gene Aliases
1700007D07Rik
Chromosome Location
Chr. 11: 70216789 - 70222794 on Build GRCm38
UniGene ID
Mm.93266
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172371.3 NP_758959.1 1, 2 514
GenBank mRNA AK028759.1 - 1, 2 515
BC039949.1 - 1, 2 302 4167719

View Full Product Details