Gene Transcripts

Gene Symbol
Nek11
Entrez Gene ID
208583
Gene Name
NIMA (never in mitosis gene a)-related expressed kinase 11
Gene Aliases
4932416N14Rik
Chromosome Location
Chr. 9: 105162156 - 105395524 on Build GRCm38
UniGene ID
Mm.347509
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172461.3 NP_766049.2 9 1181
GenBank mRNA AK030042.1 - 9 1140
AK030186.1 - 8 1057
AK043543.1 - 9 1420
AK054237.1 - 9 1075
BC119588.1 - 8 1015 40058889
BC119589.1 - 8 1015 40058890

View Full Product Details