Gene Transcripts
- Gene Symbol
- DMXL1
- Entrez Gene ID
- 1657
- Gene Name
- Dmx like 1
- Gene Aliases
- Chromosome Location
- Chr. 5: 119071002 - 119250170 on Build GRCh38
- UniGene ID
- Hs.181042
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001290321.2 |
NP_001277250.1
|
4, 5 |
855 |
|
NM_005509.5 |
NP_005500.4
|
4, 5 |
855 |
|
XM_005271909.4 |
XP_005271966.1
|
5, 6 |
553 |
|
XM_005271910.4 |
XP_005271967.1
|
4, 5 |
898 |
|
XM_005271912.2 |
XP_005271969.1
|
5, 6 |
537 |
|
XM_011543212.2 |
XP_011541514.1
|
4, 5 |
898 |
|
XM_011543213.2 |
XP_011541515.1
|
5, 6 |
483 |
|
XM_017009141.1 |
XP_016864630.1
|
5, 6 |
553 |
|
XM_017009142.1 |
XP_016864631.1
|
5, 6 |
483 |
|
XM_017009143.1 |
XP_016864632.1
|
6, 7 |
732 |
|
XM_017009144.1 |
XP_016864633.1
|
4, 5 |
435 |
|
XM_017009145.1 |
XP_016864634.1
|
4, 5 |
305 |
|
XM_017009146.1 |
XP_016864635.1
|
4, 5 |
1747 |
|
XM_017009147.1 |
XP_016864636.1
|
4, 5 |
898 |
|
GenBank mRNA |
AJ005821.1 |
- |
4, 5 |
452 |
|
BC028004.1 |
- |
5, 6 |
440 |
|
BC144537.1 |
- |
4, 5 |
398 |
9053064 |
BC150616.1 |
- |
4, 5 |
398 |
9056986 |
View Full Product Details