Gene Transcripts

Gene Symbol
Pglyrp2
Entrez Gene ID
57757
Gene Name
peptidoglycan recognition protein 2
Gene Aliases
C730002N09Rik, PGRP-L, Pglyrpl, tagL
Chromosome Location
Chr. 17: 32412459 - 32424183 on Build GRCm38
UniGene ID
Mm.86752
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271476.1 NP_001258405.1 3 457
NM_001271477.1 NP_001258406.1 2 349
NM_001271478.1 NP_001258407.1 3 457
NM_001271479.1 NP_001258408.1 2 349
NM_021319.5 NP_067294.2 2 349
GenBank mRNA AB084140.1 - 3 457
AB084141.1 - 3 457
AF149837.1 - 2 349
AF149838.1 - 2 349
AF315578.1 - 2 349
AF315584.1 - 2 349
AF317548.1 - 2 349
AF392055.1 - 3 383
AK039953.1 - 3 457
AK050019.1 - 2 350
AY282722.1 - 2 287
BC019396.1 - 2 322 5124925
EF204951.1 - 2 237

View Full Product Details