Gene Transcripts

Gene Symbol
Rpp40
Entrez Gene ID
208366
Gene Name
ribonuclease P 40 subunit
Gene Aliases
D8Bwg1265e, Rnasep1
Chromosome Location
Chr. 13: 35895104 - 35911565 on Build GRCm38
UniGene ID
Mm.35545
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145938.4 NP_666050.3 8 1763
GenBank mRNA AK048810.1 - 1763

View Full Product Details