Gene Transcripts

Gene Symbol
Mex3d
Entrez Gene ID
237400
Gene Name
mex3 homolog D (C. elegans)
Gene Aliases
BC059858, Rkhd1
Chromosome Location
Chr. 10: 80380355 - 80387651 on Build GRCm38
UniGene ID
Mm.268389
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198615.2 NP_941017.2 2 1288
GenBank mRNA AK149801.1 - 2 816
BC059858.1 - 3 408

View Full Product Details