Gene Transcripts

Gene Symbol
CFHR2
Entrez Gene ID
3080
Gene Name
complement factor H related 2
Gene Aliases
CFHL2, FHR2, HFL3
Chromosome Location
Chr. 1: 196943752 - 196959328 on Build GRCh38
UniGene ID
Hs.632450
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001312672.1 NP_001299601.1 2 304
NM_005666.3 NP_005657.1 4 676
XM_005245113.3 XP_005245170.1 3 495
XM_011509458.2 XP_011507760.1 4 648
XM_011509459.2 XP_011507761.1 4 710
XM_011509460.2 XP_011507762.1 3 485
XM_017001109.1 XP_016856598.1 3 483
GenBank mRNA AX772812.1 - 2 214
BC022283.1 - 4 665 4734331
BG563114.1 - 4 674
BG566607.1 - 2 292
X64877.1 - 4 640

View Full Product Details